missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TRUB1 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 12520-1-AP-150UL
This item is not returnable.
View return policy
Description
Pseudouridine is an abundant component of rRNAs and tRNAs and is enzymatically generated by isomerization of uridine by pseudouridine synthase (Zucchini et al., 2003 ).Specifications
TRUB1 | |
Polyclonal | |
Unconjugated | |
TRUB1 | |
PUS4, TRUB1 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
142940, 72133 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
0.13 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q8C0D0, Q8WWH5 | |
Trub1 | |
TRUB1 Fusion Protein Ag3213 | |
150 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
TRUB1 Rabbit anti-Human, Mouse, Polyclonal, Proteintech > 150 μL; Unconjugated
Spot an opportunity for improvement?Share a Content Correction