missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TRUB2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 19891-1-AP-150UL
This item is not returnable.
View return policy
Description
Pseudouridine is an abundant component of rRNAs and tRNAs and is enzymatically generated by isomerization of uridine by pseudouridine synthase.Specifications
TRUB2 | |
Polyclonal | |
Unconjugated | |
TRUB2 | |
CLONE24922, RP11 339B21.1, TRUB2 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
227682, 26995, 366012 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.18 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
O95900, Q5XFW2, Q91WG3 | |
Trub2 | |
TRUB2 Fusion Protein Ag13726 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Correzione del contenuto del prodotto
Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.
Titolo del prodotto
TRUB2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech > 150 μL; Unconjugated
Individuate un'opportunità di miglioramento?Condividi una correzione di contenuto