missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TSH-Receptor, A-Chain Mouse, Clone: TSHRA/1402, enQuireBio™
Mouse Monoclonal Antibody
2050.00 SEK - 5965.00 SEK
Specifications
Antigen | TSH-Receptor, A-Chain |
---|---|
Clone | TSHRA/1402 |
Concentration | 200ÎĽg/mL |
Applications | Immunohistochemistry |
Classification | Monoclonal |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15931028
|
EnquireBio
7253-MSM2-P0 |
20 ÎĽg | N/A | ||||||
15941028
|
EnquireBio
7253-MSM2-P1 |
100 ÎĽg |
5965.00 SEK
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Specifications
TSH-Receptor, A-Chain | |
200ÎĽg/mL | |
Monoclonal | |
Mouse | |
10mM PBS with 0.05% BSA and 0.05% Azide | |
7253 | |
Recombinant human TSHR, A-Chain protein | |
Primary | |
Short term: Store at 2-4°C. Long term, aliquot and store at -20°C. | |
62kDa |
TSHRA/1402 | |
Immunohistochemistry | |
Unconjugated | |
RUO | |
XP_005268094 | |
TSHR | |
IgG Îş | |
Protein A/G Purified | |
Cytoplasmic | |
293T Cells. Thyroid gland |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
TSH-Receptor, A-Chain Mouse, Clone: TSHRA/1402, enQuireBio™