missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TSKU Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 12370-1-AP-150UL
This item is not returnable.
View return policy
Description
The function of this protein remains unknown.Specifications
TSKU | |
Polyclonal | |
Unconjugated | |
TSKU | |
E2 induced gene 4 protein, E2IG4, LRRC54, TSK, TSKU, Tsukushi, tsukushin | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
244152, 25987 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.33 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q8CBR6, Q8WUA8 | |
Tsku | |
TSKU Fusion Protein Ag3054 | |
150 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
TSKU Rabbit anti-Human, Mouse, Polyclonal, Proteintech > 150 μL; Unconjugated
Spot an opportunity for improvement?Share a Content Correction