missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TTC31 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 21693-1-AP-150UL
This item is not returnable.
View return policy
Specifications
TTC31 | |
Polyclonal | |
Unconjugated | |
TTC31 | |
TPR repeat protein 31, TTC31 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
64427 | |
-20°C | |
Liquid |
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
0.23 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q49AM3 | |
TTC31 | |
TTC31 Fusion Protein Ag16281 | |
150 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
TTC31 Rabbit anti-Human, Polyclonal, Proteintech > 150 μL; Unconjugated
Spot an opportunity for improvement?Share a Content Correction