missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TTC39A Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 21323-1-AP-150UL
This item is not returnable.
View return policy
Description
TTC39A is a protein coding gene.Specifications
TTC39A | |
Polyclonal | |
Unconjugated | |
TTC39A | |
C1orf34, DEME 6, KIAA0452, TPR repeat protein 39A, TTC39A | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
22996, 230603, 298366 | |
-20°C | |
Liquid |
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
0.2 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
A2ACP1, Q5SRH9 | |
TTC39A | |
TTC39A Fusion Protein Ag14874 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
TTC39A Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech > 150 μL; Unconjugated
Spot an opportunity for improvement?Share a Content Correction