Learn More
TUBG2 Rabbit, anti-Human, Unconjugated, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
1645.54 SEK - 4652.90 SEK
Specifications
Antigen | TUBG2 |
---|---|
Concentration | 750 ÎĽg/mL |
Applications | Western Blot, ELISA |
Classification | Polyclonal |
Conjugate | Unconjugated |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
17231396
|
Proteintech
28009-1-AP-20UL |
20 ÎĽL |
1645.54 SEK
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
17221396
|
Proteintech
28009-1-AP-150UL |
150 ÎĽL |
4652.90 SEK
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
The TUBG2 antibody from Proteintech is a rabbit polyclonal antibody to a recombinant protein of human TUBG2. This antibody recognizes human, mouse, rat antigen. The TUBG2 antibody has been validated for the following applications: WB,ELISA analysis.
Specifications
TUBG2 | |
Western Blot, ELISA | |
Unconjugated | |
Rabbit | |
27175 |
750 ÎĽg/mL | |
Polyclonal | |
Liquid | |
Human, Mouse, Rat | |
Primary |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.