missing translation for 'onlineSavingsMsg'
Läs mer

Tubulin-α Ab-2 Mouse Monoclonal Antibody, Epredia™

Provide accurate, reproducible results with the Epredia™ Tubulin-α Ab-2, Mouse Monoclonal Antibody.

1650.00 SEK - 7070.00 SEK

Specifikationer

Antigen Tubulin, alpha Ab-2
Klona DM1A
Användningsområden Immunoassay, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
Klassificering Monoclonal
Konjugera Unconjugated
Se fler specifikationer

Produkter 6
Produktkod Brand Kvantitet Form Koncentration Pris Kvantitet och tillgänglighet  
Produktkod Brand Kvantitet Form Koncentration Pris Kvantitet och tillgänglighet  
12641667
Visa dokument
Epredia
MS-581-P0
100 μL Concentrated 200μg/mL This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Ej tillämpligt Se alternativa produkter
Spara upp till 
»
12661667
Visa dokument
Epredia
MS-581-P1ABX
100 μL Concentrated 1mg/mL This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Ej tillämpligt Se alternativa produkter
Spara upp till 
»
12671667
Visa dokument
Epredia
MS-581-PABX
200 μL Concentrated 1mg/mL This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Ej tillämpligt Se alternativa produkter
Spara upp till 
»
12624269
Visa dokument
Epredia
MS-581-P1
500 μL Concentrated 200μg/mL
ONLINE EXCLUSIVE PRICE
4595.00 SEK
4145.75 SEK / 500µL
Save 449.25 SEK
9% Av
Se alternativa produkter
Spara upp till 
»
Vänligen för att beställa denna produkten. Behöver du ett Webbkonto? Registrera ditt konto idag!
12631667
Visa dokument
Epredia
MS-581-P
1 mL Concentrated 200μg/mL This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Ej tillämpligt Se alternativa produkter
Spara upp till 
»
12601677
Visa dokument
Epredia
MS-581-R7
7 mL Prediluted Prediluted This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Ej tillämpligt Se alternativa produkter
Spara upp till 
»
Beskrivning

Beskrivning

Microtubules, the major cytoskeletal elements found in all eukaryotic cells, consist of Tubulin, which is a dimer of two 55kDa subunits: α and Β. Microtubules play key roles in chromosome segregation in mitosis, intracellular transport, ciliary and flagellar bending, and structural support of the cytoskeleton.

Host Species: Mouse

Clone: DM1A

Isotype: IgG1/κ

Species Reactivity: Human, Cow, Pig, Rat, Mouse, Guinea Pig, Gerbil, Frog and Chicken. Others not known.

Epitope: aa 426-450

Immunogen: Native chick brain microtubules

Molecular Weight: 57kDa

Positive Control: Ls174T, MAD109 cells; skin or lung

Cellular Localization: Cytoplasmic Ab-2 causes the 10nm filaments to collapse into large lateral aggregates collecting in the cell periphery or tight juxtanuclear caps. It does not block microtubule assembly. It does not inhibit polymerization or depolymerization of platelet tubulin in vitro. It blocks (by 70-80%) the ability of tubulin dimers (with GppNHp bound) to promote a stable inhibition of adenylyl cyclase.

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)
  • Western Blotting
  • Immunofluorescence
  • Immunoprecipitation (Native and denatured)
  • Electron Microscopy

Note:

USA: RUO; Int'l: RUO

Specifikationer

Specifikationer

Tubulin, alpha Ab-2
Immunoassay, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
Unconjugated
RUO
Bovine, Chicken, Frog, Gerbil, Guinea Pig, Human, Mouse, Pig, Rat
IgG2a κ
DM1A
Monoclonal
Mouse
Cytoskeletal Protein
Native chick brain microtubules
Primary
Filmer
SDS
Dokument

Dokument

Produktspecifikationer
Korrigering av produktinnehåll

Din input är viktig för oss. Fyll i det här formuläret för att ge feedback relaterad till innehållet på denna produkt.

Produkttitel

Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.

Tack! Din feedback har skickats. Fisher Scientific arbetar alltid för att förbättra vårt innehåll åt dig. Vi uppskattar din feedback.