Learn More
TUSC3 Mouse anti-Human, Unconjugated, Clone: 1D3G12, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 67382-1-IG-150UL
Description
The TUSC3 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human TUSC3. This antibody recognizes human antigen. The TUSC3 antibody has been validated for the following applications: WB, ELISA analysis.
Specifications
TUSC3 | |
Monoclonal | |
1000 μg/mL | |
Mouse | |
Primary | |
Human |
Western Blot, ELISA | |
1D3G12 | |
Unconjugated | |
150 μL | |
7991 | |
IgG1 |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.