Learn More
TUSC5 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 21556-1-AP-150UL
 View more versions of this product
Product Code. 16881145
  • 4652.90 SEK / 150µL
Description
TUSC5 (tumor suppressor candidate 5), also known as protein located at seventeen-p-thirteen point three 1, LOST1 or IFITMD3 (interferon-induced transmembrane domain-containing protein D3), is a 177 amino acid multi-pass membrane protein that belongs to the CD225 family. Thought to play a role in fat metabolism, TUSC5 is highly expressed in mammary gland, heart, smooth muscle, skeletal muscle and stomach, with lower levels found in lung and brain.
Specifications
Specifications
TUSC5
Polyclonal
Unconjugated
TUSC5
IFITMD3, LOST1, tumor suppressor candidate 5, TUSC5
Rabbit
Antigen Affinity Chromatography
RUO
286753
-20°C
Liquid
Immunohistochemistry (Paraffin), Western Blot
0.36 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Q8IXB3
TUSC5
TUSC5 Fusion Protein Ag15572
150 μL
Primary
Human
Antibody
IgG
Certificates
Promotions
Promotions
Product Content Correction
Product Title
TUSC5 Rabbit anti-Human, Polyclonal, Proteintech > 150 μL; Unconjugated