missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TWF1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 11732-1-AP-150UL
Additional Details : Weight : 0.00010kg
Description
This gene encodes twinfilin, an actin monomer-binding protein conserved from yeast to mammals. Studies of the mouse counterpart suggest that this protein may be an actin monomer-binding protein, and its localization to cortical G-actin-rich structures may be regulated by the small GTPase RAC1.Specifications
TWF1 | |
Polyclonal | |
Unconjugated | |
TWF1 | |
A6, Protein A6, Protein tyrosine kinase 9, PTK9, TWF1, Twinfilin 1 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
19230, 315265, 5756 | |
-20°C | |
Liquid |
Western Blot, Immunohistochemistry (Paraffin) | |
0.13 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q12792, Q5RJR2, Q91YR1 | |
Twf1 | |
TWF1 Fusion Protein Ag2332 | |
150 μL | |
Primary | |
Rat, Mouse, Human | |
Antibody | |
IgG |