missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TWIST2 Mouse anti-Human, Mouse, Rat, Clone: 1B12D11, Proteintech
Mouse Monoclonal Antibody
1645.54 SEK - 4652.90 SEK
Specifications
Antigen | TWIST2 |
---|---|
Clone | 1B12D11 |
Concentration | 1 mg/mL |
Applications | Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry |
Classification | Monoclonal |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16888515
|
Proteintech
66544-1-IG-20UL |
20 ÎĽL |
1645.54 SEK
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16878515
|
Proteintech
66544-1-IG-150UL |
150 ÎĽL |
4652.90 SEK
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Binds to the E-box consensus sequence 5′-CANNTG-3′ as a heterodimer and inhibits transcriptional activation by MYOD1, MYOG, MEF2A and MEF2C. Also represses expression of proinflammatory cytokines such as TNFA and IL1B. Involved in postnatal glycogen storage and energy metabolism. Inhibits the premature or ectopic differentiation of preosteoblast cells during osteogenesis, possibly by changing the internal signal transduction response of osteoblasts to external growth factors.Specifications
TWIST2 | |
1 mg/mL | |
Monoclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
bHLHa39, Dermis expressed protein 1, Dermo 1, DERMO1, twist homolog 2 (Drosophila), Twist related protein 2, TWIST2 | |
Twist2 | |
IgG1 | |
Protein G | |
Antibody |
1B12D11 | |
Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
Unconjugated | |
Mouse | |
Mouse, Human, Rat | |
P97831, Q8WVJ9, Q9D030 | |
117581, 13345, 59327 | |
TWIST2 Fusion Protein Ag2348 | |
Primary | |
-20°C | |
TWIST2 |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
TWIST2 Mouse anti-Human, Mouse, Rat, Clone: 1B12D11, Proteintech