Learn More
TXNIP Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 18243-1-AP-150UL
This item is not returnable.
View return policy
Description
The members of the zinc fingers and homeoboxes gene family are nuclear homodimeric transcriptional repressors that interact with the A subunit of nuclear factor-Y (NF-YA) and contain two C2H2-type zinc fingers and five homeobox DNA-binding domains. This gene encodes member 2 of this gene family. In addition to forming homodimers, this protein heterodimerizes with member 1 of the zinc fingers and homeoboxes family.Specifications
TXNIP | |
Polyclonal | |
Unconjugated | |
TXNIP | |
EST01027, HHCPA78, THIF, Thioredoxin binding protein 2, TXNIP, VDUP1 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
10628, 56338 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.21 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q8BG60, Q9H3M7 | |
Txnip | |
TXNIP Fusion Protein Ag13009 | |
150 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
Product Content Correction
Product Title
TXNIP Rabbit anti-Human, Mouse, Polyclonal, Proteintech > 150 μL; Unconjugated
Spot an opportunity for improvement?Übermitteln Sie eine inhaltliche Korrektur