Learn More
Justrite™ Type II AccuFlow™ Steel Safety Can
Brand: Justrite™ 7250230Z
This item is not returnable.
View return policy
Specifications
19 L | |
298 mm | |
445 mm |
JTT001 International Catalogue May 2010
File size: 6.4 MB
Product Content Correction
Product Title
Justrite™ Type II AccuFlow™ Steel Safety Can > Capacity: 19L
Spot an opportunity for improvement?Share a Content Correction