missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals [Tyr8] Atrial Natriuretic Peptide (5-27), rat

For cellular and molecular biology applications

7182.00 SEK - 12785.00 SEK

Specifications

Species Rat
Purification Method HPLC
Content And Storage -30°C to -10°C
Format Lyophilized Powder
Molecular Weight (g/mol) 2402.62
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15427819
View Documents
Thermo Scientific Chemicals
J66452.#0
5 mg This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
15437819
View Documents
Thermo Scientific Chemicals
J66452.MA
10 mg
12785.00 SEK
10mg
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Rat
-30°C to -10°C
2402.62
RUO - research use only
HPLC
Lyophilized Powder
[Tyr8] Atrial Natriuretic Peptide (5-27), rat
[Tyr8] Atriopeptin II, rat
Videos
SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Thermo Scientific Chemicals [Tyr8] Atrial Natriuretic Peptide (5-27), rat

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.