missing translation for 'onlineSavingsMsg'
Learn More
Learn More
UBE2S Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
1645.54 SEK - 4652.90 SEK
Specifications
Antigen | UBE2S |
---|---|
Concentration | 0.29 mg/mL |
Applications | Western Blot, Immunohistochemistry (Paraffin), Immunoprecipitation, Flow Cytometry |
Classification | Polyclonal |
Conjugate | Unconjugated |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16892604
|
Proteintech
14115-1-AP-20UL |
20 ÎĽL |
1645.54 SEK
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16882604
|
Proteintech
14115-1-AP-150UL |
150 ÎĽL |
4652.90 SEK
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This gene encodes a member of the ubiquitin-conjugating enzyme family. The encoded protein is able to form a thiol ester linkage with ubiquitin in a ubiquitin activating enzyme-dependent manner, a characteristic property of ubiquitin carrier proteins.Specifications
UBE2S | |
Western Blot, Immunohistochemistry (Paraffin), Immunoprecipitation, Flow Cytometry | |
Unconjugated | |
Rabbit | |
Human, Mouse, Rat | |
B5DFI8, Q16763, Q921J4 | |
27338, 292588, 77891 | |
UBE2S Fusion Protein Ag5264 | |
Primary | |
-20°C | |
UBE2S |
0.29 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
E2 EPF, E2EPF, EPF5, UBE2S, Ubiquitin carrier protein S, Ubiquitin protein ligase S | |
UBE2S | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
UBE2S Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech