missing translation for 'onlineSavingsMsg'
Learn More

UBE4B Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

Brand:  Proteintech 18148-1-AP-150UL

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16888194

  • 3313.85 SEK / 150µL
Estimated Shipment: 31-01-2022

Please to purchase this item. Need a web account? Register with us today!


Description

Description

The modification of proteins with ubiquitin is an important cellular mechanism for targeting abnormal or short-lived proteins for degradation. Ubiquitination involves at least three classes of enzymes: ubiquitin-activating enzymes, or E1s, ubiquitin-conjugating enzymes, or E2s, and ubiquitin-protein ligases, or E3s. UBE4B represents an additional conjugation factor, E4, which is involved in multiubiquitin chain assembly. UBE4B also the strongest candidate among the neuroblastoma tumor suppressors.
Specifications

Specifications

UBE4B
0.25 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
O95155, Q9ES00
E4, HDNB1, KIAA0684, UBE4B, UBOX3, UFD2
Rabbit
IgG
150 μL
-20°C
Primary
10277, 298652, 63958
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
Unconjugated
UBE4B
Liquid
Ube4b
UBE4B Fusion Protein Ag12836
Antigen Affinity Chromatography
RUO
Antibody
Polyclonal
Human, Mouse, Rat
Videos
SDS
Documents
Certificates
Special Offers

Special Offers