missing translation for 'onlineSavingsMsg'
Learn More
Learn More
UBE4B Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
1656.73 SEK - 4721.67 SEK
Specifications
Antigen | UBE4B |
---|---|
Concentration | 0.25 mg/mL |
Applications | Immunoprecipitation, Immunohistochemistry (Paraffin), Immunocytochemistry, Western Blot, Immunofluorescence |
Classification | Polyclonal |
Conjugate | Unconjugated |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16898194
|
Proteintech
18148-1-AP-20UL |
20 μL |
1656.73 SEK
20µL |
Estimated Shipment: 03-04-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16888194
|
Proteintech
18148-1-AP-150UL |
150 μL |
4721.67 SEK
150µL |
Estimated Shipment: 03-04-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
The modification of proteins with ubiquitin is an important cellular mechanism for targeting abnormal or short-lived proteins for degradation. Ubiquitination involves at least three classes of enzymes: ubiquitin-activating enzymes, or E1s, ubiquitin-conjugating enzymes, or E2s, and ubiquitin-protein ligases, or E3s. UBE4B represents an additional conjugation factor, E4, which is involved in multiubiquitin chain assembly. UBE4B also the strongest candidate among the neuroblastoma tumor suppressors.Specifications
UBE4B | |
Immunoprecipitation, Immunohistochemistry (Paraffin), Immunocytochemistry, Western Blot, Immunofluorescence | |
Unconjugated | |
Rabbit | |
Rat, Mouse, Human | |
O95155, Q9ES00 | |
10277, 298652, 63958 | |
UBE4B Fusion Protein Ag12836 | |
Primary | |
-20°C | |
UBE4B |
0.25 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
E4, HDNB1, KIAA0684, UBE4B, UBOX3, UFD2 | |
Ube4b | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |