Learn More
UBP1 Mouse anti-Human, Unconjugated, Clone: 1B10H3, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 67318-1-IG-150UL
Description
The UBP1 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human UBP1. This antibody recognizes human antigen. The UBP1 antibody has been validated for the following applications: WB, ELISA analysis.
Specifications
UBP1 | |
Monoclonal | |
1000 μg/mL | |
Mouse | |
Primary | |
Human |
Western Blot, ELISA | |
1B10H3 | |
Unconjugated | |
150 μL | |
7342 | |
IgG1 |