Learn More
UBR7 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 16903-1-AP-150UL
 View more versions of this product
Product Code. 16886664
  • 4652.90 SEK / 150µL
Description
Proteintech's UBR7 antibody 16903-1-AP is able to detect UBR7 with a theoretical MW of 48 kDa.
E3 ubiquitin-protein ligase which is a component of the N-end rule pathway, recognizes and binds to proteins bearing specific amino-terminal residues that are destabilizing according to the N-end rule, leading to their ubiquitination and subsequent degradation.
Specifications
Specifications
UBR7
Polyclonal
Unconjugated
UBR7
C14orf130, N recognin 7, UBR7
Rabbit
Antigen Affinity Chromatography
RUO
314399, 55148, 66622
-20°C
Liquid
Western Blot
0.3 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Q8BU04, Q8N806
Ubr7
UBR7 Fusion Protein Ag10445
150 μL
Primary
Human, Mouse, Rat
Antibody
IgG
Certificates
Promotions
Promotions
Product Content Correction
Product Title
UBR7 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech > 150 μL; Unconjugated