missing translation for 'onlineSavingsMsg'
Learn More
Learn More
UFSP2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 16999-1-AP-150UL
This item is not returnable.
View return policy
Description
The MW of this protein is 53 kDa, and this antibody specifically recognizes the 53 kDa protein.
Ubiquitin-fold modifier 1 (Ufm1)-specific protease 2 (UfSP2) is a cysteine protease that is responsible for the release of Ufm1 from Ufm1-conjugated cellular proteins, as well as for the generation of mature Ufm1 from its precursor.Specifications
UFSP2 | |
Polyclonal | |
Unconjugated | |
UFSP2 | |
C4orf20, UFM1 peptidase 2, Ufm1 specific protease 2, UFSP2 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
192169, 361151, 55325 | |
-20°C | |
Liquid |
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
0.19 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q5XIB4, Q99K23, Q9NUQ7 | |
Ufsp2 | |
UFSP2 Fusion Protein Ag10692 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
UFSP2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech > 150 μL; Unconjugated
Spot an opportunity for improvement?Share a Content Correction