Learn More
ULBP2 Mouse anti-Human, Unconjugated, Clone: 1A9G8, Proteintech
Mouse Monoclonal Antibody
Brand:  Proteintech 67249-1-IG-150UL
Additional Details : Weight : 0.00010kg
 View more versions of this product
Product Code. 16839775
  • 4721.67 SEK / 150µL
Estimated Shipment: 19-04-2024
to see stock.
Description
Description
The ULBP2 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human ULBP2. This antibody recognizes human antigen. The ULBP2 antibody has been validated for the following applications: WB, ELISA analysis.
Specifications
Specifications
ULBP2
Monoclonal
1000 μg/mL
Mouse
Primary
Human
Western Blot, ELISA
1A9G8
Unconjugated
150 μL
80328
IgG1