missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ULBP2 Mouse anti-Human, Unconjugated, Clone: 1A9G8, Proteintech
Mouse Monoclonal Antibody
1656.73 SEK - 4721.67 SEK
Specifications
Antigen | ULBP2 |
---|---|
Clone | 1A9G8 |
Concentration | 1000 μg/mL |
Applications | Western Blot, ELISA |
Classification | Monoclonal |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16849775
|
Proteintech
67249-1-IG-20UL |
20 μL |
1656.73 SEK
20µL |
Estimated Shipment: 14-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16839775
|
Proteintech
67249-1-IG-150UL |
150 μL |
4721.67 SEK
150µL |
Estimated Shipment: 14-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
The ULBP2 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human ULBP2. This antibody recognizes human antigen. The ULBP2 antibody has been validated for the following applications: WB, ELISA analysis.
Specifications
ULBP2 | |
1000 μg/mL | |
Monoclonal | |
Mouse | |
80328 | |
Primary |
1A9G8 | |
Western Blot, ELISA | |
Unconjugated | |
Human | |
IgG1 |