Learn More
ULBP2 Mouse anti-Human, Unconjugated, Clone: 1A9G8, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 67249-1-IG-20UL
Additional Details : Weight : 0.00010kg
Description
The ULBP2 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human ULBP2. This antibody recognizes human antigen. The ULBP2 antibody has been validated for the following applications: WB, ELISA analysis.
Specifications
ULBP2 | |
Monoclonal | |
1000 μg/mL | |
Mouse | |
Primary | |
Human |
Western Blot, ELISA | |
1A9G8 | |
Unconjugated | |
20 μL | |
80328 | |
IgG1 |