Learn More
Marienfeld Superior™ UniMark™ Uncoated Microscope Slides with Printed Marking Area Product Code.: 10462288
Estimated Shipment: 04-08-2025
Only null left
Add to basket
Marienfeld Superior™ UniMark™ Uncoated Microscope Slides with Printed Marking Area
Product Code. p-7144732
Green, Cut, Pack of 2500
Click to view available options
Color:
Green
Edges:
Cut
Unit Size:
Pack of 2500
Product Code. 10462288
Brand: Marienfeld Superior™ 0703210
Only null left
Estimated Shipment: 04-08-2025
Add to basket
Only null left
Add to basket
Marienfeld Superior™ UniMark™ miscroscope slides have a printed area which can take notes printed by most types of printers and can be written with permanent markers. Different colors of the labelling area offer the possibility to distinct the preparations (by users, etc.)
  • Made of soda lime glass of third hydrolytic class
  • In compliance with DIN ISO 8037/1
  • Pre-cleaned, ready for use, autoclavable
  • With printed marking area in different colors
  • 50 pieces in a box, 50 boxes in a carton
  • For in-vitro diagnostic (IVD) applications according to IVD directive 98/79/EC, with CE-mark, recommended best-before-date and batch number for comprehensive information and traceability
Specifications
Length (Metric) 76 mm
Width (Metric) 26 mm
No. per Case 2500
No. per Pack 50
Edges Cut
Frosted Yes
Material Soda Lime Glass Type III, Hydrolytic Class
Precleaned Yes
Thickness (Metric) 1 mm
Type Microscope Slide
Color Green
Certifications/Compliance CE Mark, DIN ISO 8037/1
Corner Style 90°
Description UniMark™ Microscope Slides with printed marking area
Quantity 50 Pieces in Cardboard Box
Dimensions (L x W) 76 x 26 mm
Show More Show Less
Product Content Correction
Product Title
Marienfeld Superior™ UniMark™ Uncoated Microscope Slides with Printed Marking Area >