Learn More
Simport™ Scientific UNISETTE™ Biopsy Processing/Embedding Cassette
Secure specimens during processing with UNISETTE™ Biopsy Processing/Embedding Cassette. The snap latch and hinge-lock design of the UNISETTE™ allow for one-hand operation and always relock securely without danger of specimen loss.
Specifications
Writing Face Angle 35°
Material Acetal
View More Specs
Description
Description
  • 35° Anterior writing area
  • One-piece construction with hinge-lock design prevent early separation of base and lid
  • Snap latch and hinge-lock design - cassettes will always relock securely
  • Allows for one-hand operation
  • Suitable for automated printers
  • Resistant to the chemical action of most histological solvents
Order Info
Each case contains 3 dispenser boxes of 500 cassettes.
Certifications
CE Marked
Specifications
Specifications
35°
Acetal
Documents
Product Certifications
For in vitro diagnostic (IVD) use only.
Product Content Correction
Product Title
Simport™ Scientific UNISETTE™ Biopsy Processing/Embedding Cassette