missing translation for 'onlineSavingsMsg'
Learn More
Learn More
UQCR Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 14793-1-AP-20UL
This item is not returnable.
View return policy
Specifications
UQCR | |
Polyclonal | |
Unconjugated | |
UQCR11 | |
0710008D09Rik, Complex III subunit 10, Complex III subunit XI, Cytochrome b c1 complex subunit 10, QCR10, UQCR, UQCR11 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
10975, 66594, 690848 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin) | |
0.13 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
O14957, Q9CPX8 | |
Uqcr11 | |
UQCR Fusion Protein Ag6491 | |
20 μL | |
Primary | |
Rat, Human, Mouse | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
UQCR Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech > 20 μL; Unconjugated
Spot an opportunity for improvement?Share a Content Correction