Läs mer
Thermo Scientific™ Urapidil hydrochloride, 98+%
alpha-1-Adrenoceptor antagonist and 5-HT1A receptor agonist
1635.81 SEK - 2336.86 SEK
Kemiska identifierare
| Analysera | ≥98% |
|---|---|
| CAS | 64887-14-5 |
| Formel vikt | 423.93g/mol |
| Synonym | 6-(3-[4-(2-Methoxypenyl)-1-piperaziny]propylamino)-1,3-dimethyluracil hydrochloride |
| Formatera | Powder |
| För användning med (applikation) | alpha-1-Adrenoceptor antagonist and 5-HT1A receptor agonist |
| Beskrivning | White |
| Produktkod | Brand | Kvantitet | Pris | Kvantitet och tillgänglighet | |||||
|---|---|---|---|---|---|---|---|---|---|
| Produktkod | Brand | Kvantitet | Pris | Kvantitet och tillgänglighet | |||||
|
15415479
|
Thermo Scientific™
J63219.03 |
1 g |
2336.86 SEK
1g |
Vänligen Logga in för att beställa denna produkten. Behöver du ett Webbkonto? Registrera ditt konto idag! | |||||
|
15435479
|
Thermo Scientific™
J63219.ME |
500 mg |
1635.81 SEK
500mg |
Vänligen Logga in för att beställa denna produkten. Behöver du ett Webbkonto? Registrera ditt konto idag! | |||||
Beskrivning
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Kemiska identifierare
| ≥98% | |
| 423.93g/mol | |
| Powder | |
| White |
| 64887-14-5 | |
| 6-(3-[4-(2-Methoxypenyl)-1-piperaziny]propylamino)-1,3-dimethyluracil hydrochloride | |
| alpha-1-Adrenoceptor antagonist and 5-HT1A receptor agonist |
Specifications
| White | |
| alpha-1-Adrenoceptor antagonist and 5-HT1A receptor agonist | |
| 64887-14-5 | |
| 423.93g/mol |
| Powder | |
| ≥98% | |
| C20H29N5O3·HCl | |
| 6-(3-[4-(2-Methoxypenyl)-1-piperaziny]propylamino)-1,3-dimethyluracil hydrochloride |
Safety and Handling
Recommended Storage : Ambient temperatures
RUO – Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.