Learn More
URB2 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 24881-1-AP-20UL
 View more versions of this product
Product Code. 16883385
  • 1645.54 SEK / 20µL
Description
URB2 gene ontology annotations related to this gene include ribosome biogenesis.
Specifications
Specifications
URB2
Polyclonal
Unconjugated
URB2
KIAA0133, NET10, URB2
Rabbit
Antigen Affinity Chromatography
RUO
9816
-20°C
Liquid
Western Blot
0.37 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Q14146
URB2
URB2 Fusion Protein Ag21667
20 μL
Primary
Human
Antibody
IgG
Certificates
Promotions
Promotions
Product Content Correction
Product Title
URB2 Rabbit anti-Human, Polyclonal, Proteintech > 20 μL; Unconjugated