Learn More
URG4 Mouse anti-Human, Unconjugated, Clone: 2D5F12, Proteintech
Mouse Monoclonal Antibody
Brand:  Proteintech 67144-1-IG-20UL
 View more versions of this product
Product Code. 16889605
  • 1645.54 SEK / 20µL
This item is not returnable. View return policy
Description
Description
The URG4 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human URG4. This antibody recognizes human, mouse, rat antigen. The URG4 antibody has been validated for the following applications: IF, IHC, WB, ELISA analysis.
Specifications
Specifications
URG4
Monoclonal
1000 μg/mL
Mouse
Primary
Human, Rat, Mouse
Western Blot, Immunohistochemistry, Immunofluorescence, ELISA
2D5F12
Unconjugated
20 μL
55665
IgG2a
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
URG4 Mouse anti-Human, Unconjugated, Clone: 2D5F12, Proteintech > 20 μL