Learn More
USF1 Mouse anti-Human, Unconjugated, Clone: 1C8F1, Proteintech
Mouse Monoclonal Antibody
Brand:  Proteintech 67114-1-IG-20UL
Additional Details : Weight : 0.00010kg
 View more versions of this product
Product Code. 16889555
  • 1355.00 SEK / 20µL
Estimated Shipment: 31-03-2023
to see stock.
Description
Description
The USF1 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human USF1. This antibody recognizes human, mouse, rat antigen. The USF1 antibody has been validated for the following applications: WB, ELISA analysis.
Specifications
Specifications
USF1
Monoclonal
1000 μg/mL
Mouse
Primary
Mouse, Rat, Human
ELISA, Western Blot
1C8F1
Unconjugated
20 μL
7391
IgG1