Learn More
USF1 Mouse anti-Human, Unconjugated, Clone: 1C8F1, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 67114-1-IG-150UL
Description
The USF1 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human USF1. This antibody recognizes human, mouse, rat antigen. The USF1 antibody has been validated for the following applications: WB, ELISA analysis.
Specifications
USF1 | |
Monoclonal | |
1000 μg/mL | |
Mouse | |
Primary | |
Human, Rat, Mouse |
Western Blot, ELISA | |
1C8F1 | |
Unconjugated | |
150 μL | |
7391 | |
IgG1 |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.