missing translation for 'onlineSavingsMsg'
Learn More
Learn More
USF2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 16614-1-AP-20UL
Additional Details : Weight : 0.00010kg
Description
USF2 consists of 43kDa (USF-1) and 44kDa (USF-2) polypeptides which independently exhibit site-specific DNA binding and are members of the c-Myc-related family of regulatory factors containing helix-loop-helix domains. USF also contains a leucine repeat that is required for efficient DNA binding.Specifications
USF2 | |
Polyclonal | |
Unconjugated | |
USF2 | |
bHLHb12, FIP, FOS interacting protein, Upstream stimulatory factor 2, USF2 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
22282, 7392, 81817 | |
-20°C | |
Liquid |
Western Blot | |
0.32 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q15853, Q63665, Q64705 | |
USF2 | |
USF2 Fusion Protein Ag9866 | |
20 μL | |
Primary | |
Human, Rat, Mouse | |
Antibody | |
IgG |