missing translation for 'onlineSavingsMsg'
Learn More
Learn More
USF2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
1656.73 SEK - 4721.67 SEK
Specifications
Antigen | USF2 |
---|---|
Concentration | 0.32 mg/mL |
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16836284
|
Proteintech
16614-1-AP-20UL |
20 μL |
1656.73 SEK
20µL |
Estimated Shipment: 15-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16826284
|
Proteintech
16614-1-AP-150UL |
150 μL |
4721.67 SEK
150µL |
Estimated Shipment: 15-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
USF2 consists of 43kDa (USF-1) and 44kDa (USF-2) polypeptides which independently exhibit site-specific DNA binding and are members of the c-Myc-related family of regulatory factors containing helix-loop-helix domains. USF also contains a leucine repeat that is required for efficient DNA binding.Specifications
USF2 | |
Western Blot | |
Unconjugated | |
Rabbit | |
Human, Rat, Mouse | |
Q15853, Q63665, Q64705 | |
22282, 7392, 81817 | |
USF2 Fusion Protein Ag9866 | |
Primary | |
-20°C | |
USF2 |
0.32 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
bHLHb12, FIP, FOS interacting protein, Upstream stimulatory factor 2, USF2 | |
USF2 | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |