missing translation for 'onlineSavingsMsg'
Learn More
Learn More
USH1G Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 21936-1-AP-150UL
This item is not returnable.
View return policy
Description
This gene encodes a protein that contains three ankyrin domains, a class I PDZ-binding motif and a sterile alpha motif. The encoded protein interacts with harmonin, which is associated with Usher syndrome type 1C. This protein plays a role in the development and maintece of the auditory and visual systems and functions in the cohesion of hair bundles formed by inner ear sensory cells. Mutations in this gene are associated with Usher syndrome type 1G (USH1G).Specifications
USH1G | |
Polyclonal | |
Unconjugated | |
USH1G | |
ANKS4A, SANS, USH1G, Usher syndrome type 1G protein | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
124590 | |
-20°C | |
Liquid |
Immunoprecipitation | |
0.42 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q495M9 | |
USH1G | |
USH1G Fusion Protein Ag16571 | |
150 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
USH1G Rabbit anti-Human, Polyclonal, Proteintech > 150 μL; Unconjugated
Spot an opportunity for improvement?Share a Content Correction