Learn More
USMG5 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 17716-1-AP-150UL
Additional Details : Weight : 0.00010kg
Description
Plays a critical role in maintaining the ATP synthase population in mitochondria.Specifications
USMG5 | |
Polyclonal | |
Unconjugated | |
USMG5 | |
HCV F transactivated protein 2, HCVFTP2, USMG5 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
171069, 66477, 84833 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.18 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q78IK2, Q96IX5, Q9JJW3 | |
Usmg5 | |
USMG5 Fusion Protein Ag12086 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |