missing translation for 'onlineSavingsMsg'
Learn More
Learn More
USMG5 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
1656.73 SEK - 4721.67 SEK
Specifications
Antigen | USMG5 |
---|---|
Concentration | 0.18 mg/mL |
Applications | Immunohistochemistry (Paraffin), Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16877654
|
Proteintech
17716-1-AP-20UL |
20 μL |
1656.73 SEK
20µL |
Estimated Shipment: 10-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16867654
|
Proteintech
17716-1-AP-150UL |
150 μL |
4721.67 SEK
150µL |
Estimated Shipment: 10-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
Plays a critical role in maintaining the ATP synthase population in mitochondria.Specifications
USMG5 | |
Immunohistochemistry (Paraffin), Western Blot | |
Unconjugated | |
Rabbit | |
Human, Mouse, Rat | |
Q78IK2, Q96IX5, Q9JJW3 | |
171069, 66477, 84833 | |
USMG5 Fusion Protein Ag12086 | |
Primary | |
-20°C | |
USMG5 |
0.18 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
HCV F transactivated protein 2, HCVFTP2, USMG5 | |
Usmg5 | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |