Learn More
BRAND™ USP Measuring Flask
With individual USP conformity certificate
Brand:  BRAND™ 956945
504.44 SEK valid until 2025-12-31
Use promo code "25339" to get your promotional price.
 View more versions of this product
Product Code. 15811198
  • 625.00 SEK / Each
Only null left
Estimated Shipment: 24-07-2025
Description
  • Boro 3.3
  • To DIN EN ISO 1042
  • Calibrated to contain (TC, In)
  • With USP batch certificate
  • TRUSTED_SUSTAINABILITY
    Specifications
    Specifications
    Polypropylene
    Borosilicate Glass (3.3)
    A
    20 mL
    Ground Joint NS 10/19
    Documents
    One moment while we fetch your results.
    Certificates
    Promotions
    Promotions
    Product Content Correction
    Product Title
    BRAND™ USP Measuring Flask > Capacity (Metric): 20 mL