missing translation for 'onlineSavingsMsg'
Learn More
Learn More
USP18 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 12153-1-AP-20UL
Additional Details : Weight : 0.00010kg
Description
USP18, a member of the deubiquitinating protease family of enzymes, removes ubiquitin adducts from a broad range of protein substrates.Specifications
USP18 | |
Polyclonal | |
Unconjugated | |
USP18 | |
43 kDa ISG15 specific protease, hUBP43, ISG43, Ubl thiolesterase 18, UBP43, USP18 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
11274 | |
-20°C | |
Liquid |
Western Blot | |
0.2 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q9UMW8 | |
USP18 | |
USP18 Fusion Protein Ag2785 | |
20 μL | |
Primary | |
Human | |
Antibody | |
IgG |