missing translation for 'onlineSavingsMsg'
Learn More
Learn More
UTP14A Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 11474-1-AP-150UL
Additional Details : Weight : 0.00010kg
Description
UTP14A, a related protein, may also be required for ribosome biogenesis, but not necessarily in a male-specific manner. The gene encoding UTP14C maps to human chromosome 13, which houses over 400 genes, such as BRCA2 and RB1, and comprises nearly 4% of the human genome. As with most chromosomes, polysomy of part or all of chromosome 13 is deleterious to development and decreases the odds of survival. Trisomy 13, also known as Patau syndrome, is deadly and the few who survive past one year suffer from permanent neurologic defects, difficulty eating and vulnerability to serious respiratory infections.Specifications
UTP14A | |
Polyclonal | |
Unconjugated | |
UTP14A | |
Antigen NY CO 16, KIAA0266, NY CO 16, SDCCAG16, UTP14A | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
10813, 317579, 72554 | |
-20°C | |
Liquid |
Western Blot, Immunofluorescence, Immunocytochemistry | |
0.13 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q640M1, Q9BVJ6 | |
Utp14a | |
UTP14A Fusion Protein Ag2027 | |
150 μL | |
Primary | |
Rat, Mouse, Human | |
Antibody | |
IgG |