Learn More
UTP3 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 18886-1-AP-150UL
 View more versions of this product
Product Code. 16888714
  • 4652.90 SEK / 150µL
This item is not returnable. View return policy
Description
Description
Mediates apoptosis and actin stress fiber dissolution.
Specifications
Specifications
UTP3
Polyclonal
Unconjugated
UTP3
CRL1, CRLZ1, Disrupter of silencing SAS10, SAS10, UTP3, UTP3 homolog
Rabbit
Antigen Affinity Chromatography
RUO
305258, 57050, 65961
-20°C
Liquid
Immunocytochemistry, Immunofluorescence, Western Blot
0.24 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Q6AXX4, Q9JI13, Q9NQZ2
Utp3
UTP3 Fusion Protein Ag13454
150 μL
Primary
Human, Mouse, Rat
Antibody
IgG
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
UTP3 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech > 150 μL; Unconjugated