Learn More
UTRN Rabbit anti-Human, Unconjugated, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 27948-1-AP-20UL
Description
The UTRN antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of human UTRN. This antibody recognizes human antigen. The UTRN antibody has been validated for the following applications: WB, ELISA analysis.
Specifications
UTRN | |
Polyclonal | |
Unconjugated | |
20 μL | |
7402 | |
IgG |
Western Blot, ELISA | |
433 μg/mL | |
Rabbit | |
Primary | |
Human |