Learn More
UVP 2UV and 3UV Benchtop Transilluminators
Up to three wavelengths available in one compact, lightweight unit
30145.00 SEK
Products 1
Product Code Brand Model Price Quantity & Availability  
Product Code Brand Model Price Quantity & Availability  
11308453
View Documents
UVP
95-0417-02
LMs-20E
30145.00 SEK
Each
Only null left
Estimated Shipment: 12-08-2025
Add to basket
Description
Description
  • 8-watt models provide economical alternative to High Performance UV Transilluminators
  • All models include hinged UV blocking cover with adjustable height and UVG long-life filter
Two single-intensity options
  • 3UV models offer selection of longwave UV for extended viewing of gels without photonicking; midrange with increased fluorescence photodocumentation and nucleic acid visualization; shortwave for sample irradiation
  • 2UV models provide longwave and midrange UV
1International model.
Order Info
2UV Dimensions (including cover): 9 1/2L x 13 1/4W x 4 3/4 in.H (24 x 33.7 x 12.1cm); 3UV Dimensions (including cover): 11L x 14W x 6 in.H (28 x 35.6 x 15.2cm)
Certifications
115V models are ETL, ETL/c certified; 230V1 models are CE certified
Documents
Product Certifications
Product Content Correction
Product Title
UVP 2UV and 3UV Benchtop Transilluminators