missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Vα 11.1, 11.2[b,d] TCR Rat anti-Mouse, BV510, Clone: RR8-1, BD OptiBuild™
Rat Monoclonal Antibody
Brand: BD Biosciences 744922
4440.50 SEK valid until 2025-12-31
Use promo code "25339" to get your promotional price.
This item is not returnable.
View return policy
Description
Vα 11.1, 11.2[b,d] TCR Monoclonal antibody specifically detects Vα 11.1, 11.2[b,d] TCR in Mouse samples. It is validated for Flow Cytometry.
Specifications
Vα 11.1, 11.2[b,d] TCR | |
Monoclonal | |
0.2 mg/mL | |
Rat | |
RUO | |
Mouse | |
Purified |
Flow Cytometry | |
RR8-1 | |
Brilliant Violet 510 | |
50 ÎĽg | |
Primary | |
Refrigerate (2 to 8°C) | |
IgG2b Îş |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Vα 11.1, 11.2[b,d] TCR Rat anti-Mouse, BV510, Clone: RR8-1, BD OptiBuild™ > Ms VAlp 11.1-11.2bd TCR BV510 RR8-1 50 μg
Spot an opportunity for improvement?Share a Content Correction