Learn More
BRAND™ Polypropylene Valve Cap
For closing valve block. Suitable for autoclaving.
Brand:  BRAND™ 704552
280.23 SEK valid until 2025-08-24
Use promo code "24117" to get your promotional price.
 View more versions of this product
Product Code. 10039800
  • 346.00 SEK / Each
Genomics Powerhouse
Use promo code 24117 at checkout
View Available Promotions
Only null left
In Stock
Description
  • Polypropylene construction
  • Autoclavable
  • For use with seripettor™
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Valve Cap
Polypropylene
Seripettor™ / Seripettor™ pro
Autoclavable
2, 10 mL
Documents
Product Information
Certificates
Promotions
Promotions
Product Content Correction
Product Title
BRAND™ Polypropylene Valve Cap > Capacity 2+10mL