missing translation for 'onlineSavingsMsg'
Learn More
Learn More
VAMP3/Cellubrevin Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
1645.54 SEK - 4652.90 SEK
Especificaciones
Antigen | VAMP3/Cellubrevin |
---|---|
Concentration | 0.35 mg/mL |
Applications | Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
CĂłdigo de producto | Marca | Quantity | Precio | Cantidad y disponibilidad | |||||
---|---|---|---|---|---|---|---|---|---|
CĂłdigo de producto | Marca | Quantity | Precio | Cantidad y disponibilidad | |||||
16897253
|
Proteintech
10702-1-AP-20UL |
20 ÎĽL |
1645.54 SEK
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16887253
|
Proteintech
10702-1-AP-150UL |
150 ÎĽL |
4652.90 SEK
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
DescripciĂłn
Synaptobrevins/VAMPs, syntaxins, and the 25-kD synaptosomal-associated protein are the main components of a protein complex involved in the docking and/or fusion of synaptic vesicles with the presynaptic membrane. This gene is a member of the vesicle-associated membrane protein (VAMP)/synaptobrevin family. Because of its high homology to other known VAMPs, its broad tissue distribution, and its subcellular localization, the protein encoded by this gene was shown to be the human equivalent of the rodent cellubrevin. In platelets the protein resides on a compartment that is not mobilized to the plasma membrane on calcium or thrombin stimulation.Especificaciones
VAMP3/Cellubrevin | |
Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
Unconjugated | |
Rabbit | |
Human, Rat, Mouse | |
P63024, P63025, Q15836 | |
22319, 29528, 9341 | |
VAMP3/Cellubrevin Fusion Protein Ag1156 | |
Primary | |
-20°C | |
VAMP3 |
0.35 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
CEB, Cellubrevin, SYB3, Synaptobrevin 3, VAMP 3, VAMP3 | |
VAMP3 | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |
ÂżDetecta una oportunidad de mejora?Comparta una correcciĂłn de contenido
CorrecciĂłn del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
TĂtulo del producto
VAMP3/Cellubrevin Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech