Learn More
Vaspin Mouse anti-Human, Unconjugated, Clone: 2A1B6, Proteintech
Mouse Monoclonal Antibody
Brand:  Proteintech 67573-1-IG-150UL
 View more versions of this product
Product Code. 16890276
  • 4652.90 SEK / 150µL
This item is not returnable. View return policy
Description
Description
The Vaspin antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human Vaspin. This antibody recognizes human antigen. The Vaspin antibody has been validated for the following applications: WB, ELISA analysis.
Specifications
Specifications
Vaspin
Monoclonal
1000 μg/mL
Mouse
Primary
Human
Western Blot, ELISA
2A1B6
Unconjugated
150 μL
145264
IgG1
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Vaspin Mouse anti-Human, Unconjugated, Clone: 2A1B6, Proteintech > 150 μL