missing translation for 'onlineSavingsMsg'
Learn More
Learn More
VAV2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 21924-1-AP-150UL
This item is not returnable.
View return policy
Description
VAV2 is the second member of the VAV guanine nucleotide exchange factor family of oncogenes. Unlike VAV1, which is expressed exclusively in hematopoietic cells, VAV2 transcripts were found in most tissues. Alternatively spliced transcript variants encoding different isoforms have been found for this gene.Specifications
VAV2 | |
Polyclonal | |
Unconjugated | |
VAV2 | |
VAV 2, VAV2 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
22325, 296603, 7410 | |
-20°C | |
Liquid |
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
0.27 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
P52735, Q60992 | |
VAV2 | |
VAV2 Fusion Protein Ag16458 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
VAV2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech > 150 μL; Unconjugated
Spot an opportunity for improvement?Share a Content Correction