missing translation for 'onlineSavingsMsg'
Learn More
Learn More
VGAT Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
1645.54 SEK - 4652.90 SEK
Specifications
Antigen | VGAT |
---|---|
Concentration | 0.13 mg/mL |
Applications | Immunofluorescence, Western Blot, Immunoprecipitation, Immunocytochemistry |
Classification | Polyclonal |
Conjugate | Unconjugated |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16893124
|
Proteintech
14471-1-AP-20UL |
20 ÎĽL |
1645.54 SEK
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16883124
|
Proteintech
14471-1-AP-150UL |
150 ÎĽL |
4652.90 SEK
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
The Vesicular GABA Amino Acid Transporter (VGAT) is responsible for transport of the inhibitory neurotransmitter into synaptic vesicles (McIntire et al., 1997). The VGAT protein (also known as the Vesicular Inhibitory Amino Aid Transporter or VIAAT) is expressed in synaptic vesicles of both glycine and GABAergic synapses throughout the CNS (Chaudhry et al., 1998). Expression of the VGAT protein changes during development and also in response to patterns of neuronal activity (De et al., 2005).Specifications
VGAT | |
Immunofluorescence, Western Blot, Immunoprecipitation, Immunocytochemistry | |
Unconjugated | |
Rabbit | |
Human, Mouse, Rat | |
O35458, O35633, Q9H598 | |
140679, 22348, 83612 | |
VGAT Fusion Protein Ag5843 | |
Primary | |
-20°C | |
SLC32A1 |
0.13 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
GABA and glycine transporter, hVIAAT, SLC32A1, Vesicular GABA transporter, VGAT, VIAAT | |
Slc32a1 | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
VGAT Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech