Läs mer
Fisherbrand™ Polyethylene Sample Bags
For use in the laboratory to carry samples into a sterile environment.
Brand:  Fisherbrand™ 14955189
 Se fler versioner av denna produkt
Produktkod. 11984395
  • 278.00 SEK / Paket med 500
Bara null kvar
Beräknad leverans: 08-09-2026
Lägg i varukorgen
Beskrivning
Beskrivning
  • Secure, contaminant free bags to ensure dependable analysis results
  • Made from heavy gauge, virgin polyethylene that meets FDA specifications
  • Can accommodate solid, semi solid and liquid materials
  • Easy to use, insert sample and seal by twirling the bag away outward four times and bending wire ends inward
  • Economical and efficient way to collect, contain and carry samples